Assay Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507271.2 | XP_003507319.1 | 8 - 9 | 992 | 57 | |
| XM_007647840.1 | XP_007646030.1 | 9 - 10 | 1270 | 57 | ||
More Information
Set Membership: |
|

Dph1
Genomic Map