Assay Details
| Gene Symbol: Wnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654220.1 | XP_007652410.1 | 4 - 5 | 749 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt2b| Gene Symbol: Wnt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654220.1 | XP_007652410.1 | 4 - 5 | 749 | 74 | |
Set Membership: |
|