Assay Details
| Gene Symbol: Casp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649342.1 | XP_007647532.1 | 8 - 9 | 1089 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casp16| Gene Symbol: Casp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649342.1 | XP_007647532.1 | 8 - 9 | 1089 | 70 | |
Set Membership: |
|