Assay Details
| Gene Symbol: Znf280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007636323.1 | XP_007634513.1 | 11 - 12 | 1374 | 67 | |
| GenBank mRNA | JI889715.1 | - | 6 - 7 | 852 | 67 | |
| JP045667.1 | - | 6 - 7 | 852 | 67 | ||
More Information
Set Membership: |
|

Znf280c
Genomic Map