Assay Details
| Gene Symbol: LOC100757714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653902.1 | XP_007652092.1 | 4 - 5 | 835 | 81 | |
| XM_007653907.1 | XP_007652097.1 | 4 - 5 | 722 | 81 | ||
More Information
Set Membership: |
|

LOC100757714
Genomic Map