Assay Details
| Gene Symbol: Anapc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496894.2 | XP_003496942.1 | 2 - 3 | 173 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anapc11| Gene Symbol: Anapc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496894.2 | XP_003496942.1 | 2 - 3 | 173 | 101 | |
Set Membership: |
|