Assay Details
| Gene Symbol: Mef2bnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499118.2 | XP_003499166.1 | 2 - 3 | 478 | 100 | |
| XM_007637592.1 | XP_007635782.1 | 2 - 3 | 478 | 100 | ||
| XM_007637597.1 | XP_007635787.1 | 2 - 3 | 477 | 100 | ||
| GenBank mRNA | JI874331.1 | - | 2 - 3 | 432 | 100 | |

Mef2bnb
Genomic Map