Assay Details
| Gene Symbol: Mef2bnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499118.2 | XP_003499166.1 | 3 - 4 | 538 | 62 | |
| XM_007637592.1 | XP_007635782.1 | 3 - 4 | 538 | 62 | ||
| XM_007637597.1 | XP_007635787.1 | 3 - 4 | 537 | 62 | ||
| GenBank mRNA | JI874331.1 | - | 3 - 4 | 492 | 62 | |
More Information
Set Membership: |
|

Mef2bnb
Genomic Map