Assay Details
| Gene Symbol: Rarb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499184.2 | XP_003499232.1 | 3 - 4 | 450 | 67 | |
| XM_003499185.1 | XP_003499233.1 | 3 - 4 | 471 | 67 | ||
| XM_007639195.1 | XP_007637385.1 | 3 - 4 | 450 | 67 | ||
More Information
Set Membership: |
|

Rarb
Genomic Map