Assay Details
Gene Symbol: LOC100764544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500591.2 | XP_003500639.1 | 2 - 3 | 100 | 83 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100764544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500591.2 | XP_003500639.1 | 2 - 3 | 100 | 83 |