Assay Details
| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505544.2 | XP_003505592.1 | 6 - 7 | 821 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmx1a| Gene Symbol: Lmx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505544.2 | XP_003505592.1 | 6 - 7 | 821 | 65 | |
Set Membership: |
|