Assay Details
| Gene Symbol: Eda2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506758.2 | XP_003506806.1 | 5 - 6 | 575 | 73 | |
| XM_007647292.1 | XP_007645482.1 | 5 - 6 | 575 | 73 | ||
| XM_007647293.1 | XP_007645483.1 | 5 - 6 | 575 | 73 | ||
| XM_007647294.1 | XP_007645484.1 | 5 - 6 | 575 | 73 | ||
| XM_007647295.1 | XP_007645485.1 | 5 - 6 | 486 | 73 | ||
| XM_007647296.1 | XP_007645486.1 | 4 - 5 | 485 | 73 | ||
More Information
Set Membership: |
|

Eda2r
Genomic Map