Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509468.2 | XP_003509516.1 | 2 - 3 | 200 | 63 | |
| XM_007650058.1 | XP_007648248.1 | 2 - 3 | 200 | 63 | ||
| GenBank mRNA | JI866249.1 | - | 2 - 3 | 180 | 63 | |
More Information
Set Membership: |
|

Cby1
Genomic Map