Assay Details
| Gene Symbol: LOC100773370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895109.1 | - | 8 - 9 | 1201 | 99 | |
| JP045167.1 | - | 8 - 9 | 1201 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773370| Gene Symbol: LOC100773370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI895109.1 | - | 8 - 9 | 1201 | 99 | |
| JP045167.1 | - | 8 - 9 | 1201 | 99 | ||
Set Membership: |
|