Assay Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608059.1 | XP_007606249.1 | 1 - 2 | 200 | 90 | |
| GenBank mRNA | JI895946.1 | - | 714 | 90 | ||
More Information
Set Membership: |
|

Smarce1
Genomic Map