Assay Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643210.1 | XP_007641400.1 | 8 - 9 | 1365 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g2| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643210.1 | XP_007641400.1 | 8 - 9 | 1365 | 59 | |
Set Membership: |
|