Assay Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652886.1 | XP_007651076.1 | 4 - 5 | 453 | 96 | |
| GenBank mRNA | JI885989.1 | - | 1 - 2 | 143 | 96 | |
| JP048767.1 | - | 4 - 5 | 385 | 96 | ||
More Information
Set Membership: |
|

Stx7
Genomic Map