Assay Details
| Gene Symbol: H6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651585.1 | XP_007649775.1 | 5 - 6 | 1304 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H6pd| Gene Symbol: H6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651585.1 | XP_007649775.1 | 5 - 6 | 1304 | 68 | |
Set Membership: |
|