Assay Details
| Gene Symbol: LOC100773370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651122.1 | XP_007649312.1 | 22 - 23 | 2406 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773370| Gene Symbol: LOC100773370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651122.1 | XP_007649312.1 | 22 - 23 | 2406 | 68 | |
Set Membership: |
|