Assay Details
| Gene Symbol: LOC100773370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651122.1 | XP_007649312.1 | 29 - 30 | 3079 | 110 | |
| GenBank mRNA | JI895109.1 | - | 311 | 110 | ||
| JP045167.1 | - | 311 | 110 | |||
More Information
Set Membership: |
|

LOC100773370
Genomic Map