Assay Details
| Gene Symbol: Map1lc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495104.2 | XP_003495152.1 | 1 - 2 | 116 | 62 | |
| GenBank mRNA | JI896830.1 | - | 1 - 2 | 124 | 62 | |
| JP037804.1 | - | 1 - 2 | 124 | 62 | ||
More Information
Set Membership: |
|

Map1lc3b
Genomic Map