Assay Details
| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496664.2 | XP_003496712.1 | 2 - 3 | 239 | 69 | |
| XM_007652221.1 | XP_007650411.1 | 2 - 3 | 239 | 69 | ||
| XM_007652223.1 | XP_007650413.1 | 2 - 3 | 239 | 69 | ||
| XM_007652226.1 | XP_007650416.1 | 2 - 3 | 239 | 69 | ||
| GenBank mRNA | JI882020.1 | - | 2 - 3 | 201 | 69 | |
| JP043000.1 | - | 2 - 3 | 201 | 69 | ||
More Information
Set Membership: |
|

Cdk8
Genomic Map