Assay Details
| Gene Symbol: Cnga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498302.1 | XP_003498350.1 | 5 - 6 | 613 | 60 | |
| XM_007625619.1 | XP_007623809.1 | 6 - 7 | 622 | 60 | ||
| XM_007625634.1 | XP_007623824.1 | 4 - 5 | 499 | 60 | ||
More Information
Set Membership: |
|

Cnga3
Genomic Map