Assay Details
| Gene Symbol: Arsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500320.2 | XP_003500368.1 | 11 - 12 | 2089 | 58 | |
| XM_007641034.1 | XP_007639224.1 | 10 - 11 | 2040 | 58 | ||
| XM_007641035.1 | XP_007639225.1 | 10 - 11 | 1977 | 58 | ||
| XM_007641036.1 | XP_007639226.1 | 9 - 10 | 1780 | 58 | ||
| XM_007641037.1 | XP_007639227.1 | 10 - 11 | 1891 | 58 | ||
| XM_007641038.1 | XP_007639228.1 | 8 - 9 | 1729 | 58 | ||
| XM_007641039.1 | XP_007639229.1 | 9 - 10 | 1841 | 58 | ||
| XM_007641040.1 | XP_007639230.1 | 8 - 9 | 843 | 58 | ||
| XM_007641041.1 | XP_007639231.1 | 6 - 7 | 709 | 58 | ||
| GenBank mRNA | JI894521.1 | - | 6 - 7 | 58 | ||
More Information
Set Membership: |
|

Arsg
Genomic Map