Assay Details
| Gene Symbol: Med19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506151.2 | XP_003506199.1 | 1 - 2 | 303 | 88 | |
| GenBank mRNA | JI886597.1 | - | 1 - 2 | 60 | 88 | |
| JP043808.1 | - | 1 - 2 | 60 | 88 | ||
More Information
Set Membership: |
|

Med19
Genomic Map