Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509901.2 | XP_003509949.1 | 8 - 9 | 772 | 64 | |
| XM_007650551.1 | XP_007648741.1 | 8 - 9 | 722 | 64 | ||
| XM_007650552.1 | XP_007648742.1 | 9 - 10 | 835 | 64 | ||
| XM_007650553.1 | XP_007648743.1 | 9 - 10 | 785 | 64 | ||
| GenBank mRNA | JI874444.1 | - | 6 - 7 | 609 | 64 | |
| JP050277.1 | - | 6 - 7 | 609 | 64 | ||
More Information
Set Membership: |
|

Esd
Genomic Map