Assay Details
| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511964.2 | XP_003512012.1 | 2 - 3 | 294 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp3| Gene Symbol: Aqp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511964.2 | XP_003512012.1 | 2 - 3 | 294 | 62 | |
Set Membership: |
|