Assay Details
| Gene Symbol: LOC100753922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI882440.1 | - | 2 - 3 | 232 | 92 | |
| JP041652.1 | - | 2 - 3 | 230 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100753922| Gene Symbol: LOC100753922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI882440.1 | - | 2 - 3 | 232 | 92 | |
| JP041652.1 | - | 2 - 3 | 230 | 92 | ||
Set Membership: |
|