Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652551.1 | XP_007650741.1 | 1 - 2 | 266 | 78 | |
| XM_007652552.1 | XP_007650742.1 | 1 - 2 | 262 | 78 | ||
| XM_007652553.1 | XP_007650743.1 | 1 - 2 | 264 | 78 | ||
| XM_007652554.1 | XP_007650744.1 | 1 - 2 | 261 | 78 | ||
| XM_007652555.1 | XP_007650745.1 | 1 - 2 | 261 | 78 | ||
More Information
Set Membership: |
|

Vti1a
Genomic Map