Assay Details
| Gene Symbol: Sgol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500455.2 | XP_003500503.1 | 2 - 3 | 428 | 72 | |
| GenBank mRNA | JI886452.1 | - | 2 - 3 | 428 | 72 | |
| JP052647.1 | - | 2 - 3 | 428 | 72 | ||
More Information
Set Membership: |
|

Sgol1
Genomic Map