Assay Details
| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501121.2 | XP_003501169.1 | 1 - 2 | 187 | 65 | |
| GenBank mRNA | JI882876.1 | - | 1 - 2 | 165 | 65 | |
| JP055786.1 | - | 1 - 2 | 165 | 65 | ||
More Information
Set Membership: |
|

Pxmp4
Genomic Map