Assay Details
| Gene Symbol: Fam186b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513497.1 | XP_003513545.1 | 5 - 6 | 2362 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam186b| Gene Symbol: Fam186b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513497.1 | XP_003513545.1 | 5 - 6 | 2362 | 75 | |
Set Membership: |
|