Assay Details
| Gene Symbol: LOC100762283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514013.1 | XP_003514061.1 | 13 - 14 | 1942 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100762283| Gene Symbol: LOC100762283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514013.1 | XP_003514061.1 | 13 - 14 | 1942 | 63 | |
Set Membership: |
|