Assay Details
| Gene Symbol: Stx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007655014.1 | XP_007653204.1 | 4 - 5 | 385 | 58 | |
| XM_007655016.1 | XP_007653206.1 | 4 - 5 | 388 | 58 | ||
| XM_007655017.1 | XP_007653207.1 | 4 - 5 | 517 | 58 | ||
More Information
Set Membership: |
|

Stx2
Genomic Map