Assay Details
| Gene Symbol: Tp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645115.1 | XP_007643305.1 | 1 - 2 | 1044 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tp53tg5| Gene Symbol: Tp53tg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645115.1 | XP_007643305.1 | 1 - 2 | 1044 | 78 | |
Set Membership: |
|