Assay Details
| Gene Symbol: Ndufs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497326.2 | XP_003497374.1 | 4 - 5 | 463 | 75 | |
| GenBank mRNA | JI877569.1 | - | 4 - 5 | 466 | 75 | |
| JP049803.1 | - | 4 - 5 | 466 | 75 | ||
More Information
Set Membership: |
|

Ndufs3
Genomic Map