Assay Details
Gene Symbol: Znf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500807.2 | XP_003500855.1 | 2 - 3 | 635 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
