Assay Details
| Gene Symbol: Kiaa0319l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500884.2 | XP_003500932.1 | 13 - 14 | 2294 | 60 | |
| XM_007641598.1 | XP_007639788.1 | 13 - 14 | 2211 | 60 | ||
| XM_007641599.1 | XP_007639789.1 | 14 - 15 | 2430 | 60 | ||
| XM_007641600.1 | XP_007639790.1 | 13 - 14 | 2121 | 60 | ||
| XM_007641601.1 | XP_007639791.1 | 13 - 14 | 2197 | 60 | ||
| XM_007641603.1 | XP_007639793.1 | 13 - 14 | 2152 | 60 | ||
| XM_007641604.1 | XP_007639794.1 | 13 - 14 | 2160 | 60 | ||
| XM_007641605.1 | XP_007639795.1 | 12 - 13 | 2093 | 60 | ||
| XM_007641606.1 | XP_007639796.1 | 12 - 13 | 2001 | 60 | ||
| XM_007641607.1 | XP_007639797.1 | 12 - 13 | 2001 | 60 | ||
| GenBank mRNA | JI872842.1 | - | 4 - 5 | 440 | 60 | |
More Information
Set Membership: |
|

Kiaa0319l
Genomic Map