Assay Details
| Gene Symbol: Rps3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504173.2 | XP_003504221.1 | 5 - 6 | 747 | 119 | |
| GenBank mRNA | JI885417.1 | - | 5 - 6 | 734 | 119 | |
| JP062748.1 | - | 5 - 6 | 730 | 119 | ||
More Information
Set Membership: |
|

Rps3a
Genomic Map