Assay Details
| Gene Symbol: Znf706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504325.2 | XP_003504373.1 | 1 - 2 | 141 | 83 | |
| GenBank mRNA | JI862765.1 | - | 2 - 3 | 83 | ||
| JP039381.1 | - | 2 - 3 | 324 | 83 | ||
More Information
Set Membership: |
|

Znf706
Genomic Map