Assay Details
| Gene Symbol: Nsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507990.2 | XP_003508038.1 | 5 - 6 | 833 | 79 | |
| GenBank mRNA | JI867113.1 | - | 5 - 6 | 832 | 79 | |
More Information
Set Membership: |
|

Nsa2
Genomic Map