Assay Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511231.2 | XP_003511279.1 | 4 - 5 | 1455 | 71 | |
| GenBank mRNA | JI887308.1 | - | 4 - 5 | 1455 | 71 | |
| JP047484.1 | - | 4 - 5 | 1455 | 71 | ||
More Information
Set Membership: |
|

Ubap1
Genomic Map