Assay Details
| Gene Symbol: Rpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514042.2 | XP_003514090.1 | 1 - 2 | 103 | 109 | |
| GenBank mRNA | JI882894.1 | - | 109 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl34| Gene Symbol: Rpl34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514042.2 | XP_003514090.1 | 1 - 2 | 103 | 109 | |
| GenBank mRNA | JI882894.1 | - | 109 | |||
Set Membership: |
|