Assay Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515070.2 | XP_003515118.1 | 2 - 3 | 179 | 72 | |
| XM_007655085.1 | XP_007653275.1 | 2 - 3 | 137 | 72 | ||
More Information
Set Membership: |
|

Spp1
Genomic Map