Assay Details
| Gene Symbol: Atp6v0e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500820.2 | XP_003500868.1 | 2 - 3 | 274 | 66 | |
| GenBank mRNA | JI889023.1 | - | 2 - 3 | 247 | 66 | |
More Information
Set Membership: |
|

Atp6v0e1
Genomic Map