Assay Details
| Gene Symbol: Csgalnact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504270.2 | XP_003504318.1 | 7 - 8 | 1852 | 80 | |
| XM_007644822.1 | XP_007643012.1 | 7 - 8 | 1618 | 80 | ||
| GenBank mRNA | JI882604.1 | - | 5 - 6 | 681 | 80 | |
| JP038299.1 | - | 5 - 6 | 681 | 80 | ||
More Information
Set Membership: |
|

Csgalnact2
Genomic Map