Assay Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510502.2 | XP_003510550.1 | 3 - 4 | 524 | 68 | |
| GenBank mRNA | JI895582.1 | - | 3 - 4 | 488 | 68 | |
More Information
Set Membership: |
|

Rab11b
Genomic Map