Assay Details
| Gene Symbol: LOC100773461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512565.1 | XP_003512613.1 | 7 - 8 | 1144 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773461| Gene Symbol: LOC100773461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512565.1 | XP_003512613.1 | 7 - 8 | 1144 | 65 | |
Set Membership: |
|