Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653524.1 | XP_007651714.1 | 2 - 3 | 447 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653524.1 | XP_007651714.1 | 2 - 3 | 447 | 113 |
Set Membership: |
![]() ![]() ![]() |