Assay Details
Gene Symbol: Hiat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653524.1 | XP_007651714.1 | 5 - 6 | 913 | 110 | |
GenBank mRNA | JI888543.1 | - | 3 - 4 | 853 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
